Proteintech
Manufacturer Code:165491AP150UL
Catalog # 50-173-1019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCK B CCK B receptor CCK BR CCK2 R CCK2R CCKBR CCKRB Cholecystokinin 2 receptor cholecystokinin B receptor GASR; CCK-B receptor; CCK-BR; CCK2 receptor; CCK2-R; Cholecystokinin-2 receptor; gastrin receptor; Gastrin/cholecystokinin type B receptor
Gene Aliases: CCK-B; CCK2R; CCKBR; CCKRB; GASR
UniProt ID: (Human) P32239
Entrez Gene ID: (Human) 887
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.