Invitrogen
Manufacturer Code:PA518609
Catalog # PIPA518609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KFDASQKKSAKEKR) corresponding to the internal amino acids of Cckar |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CCK-A receptor; CCK1-R; Cckar cholecystokinin A receptor AW106902; Cholecystokinin receptor type A; Cholecystokinin-1 receptor
Gene Aliases: Cck-ar; Cckar
UniProt ID: (Rat) P30551
Entrez Gene ID: (Rat) 24889
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.