Invitrogen
Manufacturer Code:PA518385
Catalog # PIPA518385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
peptide-ELISA (pep-ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (FEASQKKSAKERKPST) corresponding to the internal amino acids of CCKAR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CCK-A receptor; CCK-AR; CCK1-R; CCKAR cholecystokinin A receptor HGNC:1570 CCK-A CCK1-R CCKRA cholecystokinin type-A receptor cholecystokinin-1 receptor; Cholecystokinin receptor type A; cholecystokinin type-A receptor; Cholecystokinin-1 receptor
Gene Aliases: CCK-A; CCK1-R; CCK1R; CCKAR; CCKRA
UniProt ID: (Human) P32238
Entrez Gene ID: (Human) 886
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.