Invitrogen
Manufacturer Code:PA131121
Catalog # PIPA131121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Poly Lysin conjugated synthetic peptide: RFLLPPNDVMQQSWH corresponding to amino acids 197-210 of Human CCKAR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCK-A CCK1-R CCKRA; CCK-A receptor; CCK-AR; CCK1-R; Cholecystokinin receptor type A; cholecystokinin type-A receptor; Cholecystokinin-1 receptor
Gene Aliases: CCK-A; CCK-AR; CCK1-R; CCK1R; CCKAR; CCKRA
UniProt ID: (Human) P32238, (Guinea pig) Q63931
Entrez Gene ID: (Pig) 100623128, (Human) 886, (Guinea pig) 100719593
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.