Invitrogen
Manufacturer Code:PA136144
Catalog # 01-672-100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from the C-terminal of the rat CCK-A receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCK-A CCK1-R CCKRA; CCK-A receptor; CCK-AR; CCK1-R; Cholecystokinin receptor type A; Cholecystokinin-1 receptor
Gene Aliases: AW106902; Cck-ar; Cckar
UniProt ID: (Rat) P30551, (Mouse) O08786
Entrez Gene ID: (Rat) 24889, (Mouse) 12425
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.