R&D Systems
Manufacturer Code:AF2680SP
Catalog # AF2680SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Rat CCK-A R synthetic peptide Lys256-Glu267 Accession # P30551 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCK-A receptor; CCK-AR; CCK1-R; CCK1R CCK1-R CCK-A CCK-A R CCK-A receptor CCKAR CCK-AR CCKRA cholecystokinin A receptor cholecystokinin receptor type A cholecystokinin type-A receptor Cholecystokinin-1 receptor; Cholecystokinin receptor type A; cholecystokinin type-A receptor; Cholecystokinin-1 receptor
Gene Aliases: CCK-A; CCK1-R; CCK1R; CCKAR; CCKRA
UniProt ID: (Human) P32238
Entrez Gene ID: (Human) 886
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.