Invitrogen
Manufacturer Code:PA542641
Catalog # PIPA542641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CCDC90A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coiled-coil domain containing 90A; Coiled-coil domain-containing protein 90A, mitochondrial; formin-like protein 16; MCU regulator 1; Mitochondrial calcium uniporter regulator 1
Gene Aliases: 6230416A05Rik; AU015498; AV136929; AW554392; C6orf79; C88263; CCDC90A; FMP32; MCUR1; RGD1307673
UniProt ID: (Human) Q96AQ8, (Mouse) Q9CXD6
Entrez Gene ID: (Dog) 610095, (Human) 63933, (Horse) 100065066, (Bovine) 617129, (Mouse) 76137, (Rat) 291034
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.