Novus Biologicals
Manufacturer Code:NBP215755
Catalog # NBP215755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human CCDC90A. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf79 coiled-coil domain containing 90A coiled-coil domain-containing protein 90A mitochondrial FLJ20958; coiled-coil domain containing 90A; Coiled-coil domain-containing protein 90A, mitochondrial; MCU regulator 1; Mitochondrial calcium uniporter regulator 1
Gene Aliases: C6orf79; CCDC90A; FMP32; MCUR1
UniProt ID: (Human) Q96AQ8
Entrez Gene ID: (Human) 63933
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.