Novus Biologicals
Manufacturer Code:NBP292861100UL
Catalog # NB027587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-241 of human CCDC59 (NP_054886.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BR22HSPC128 coiled-coil domain containing 59 Coiled-coil domain-containing protein 59 FLJ10294 TAP26DKFZp686K1021 thyroid transcription factor 1-associated protein 26 TTF-1-associated protein 26 TTF-1-associated protein BR2; Coiled-coil domain-containing protein 59; Thyroid transcription factor 1-associated protein 26; TTF-1-associated protein 26; TTF-1-associated protein BR2
Gene Aliases: BR22; CCDC59; HSPC128; TAP26
UniProt ID: (Human) Q9P031
Entrez Gene ID: (Human) 29080
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.