Novus Biologicals
Manufacturer Code:NBP292173100UL
Catalog # NB026237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-106 of human COA3 (NP_001035521.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coiled-coil domain containing 56 coiled-coil domain-containing protein 56 FLJ50764 HSPC009; Coiled-coil domain-containing protein 56; Cytochrome c oxidase assembly factor 3 homolog, mitochondrial; cytochrome c oxidase assembly protein 3 homolog, mitochondrial; Mitochondrial translation regulation assembly intermediate of cytochrome c oxidase protein of 12 kDa
Gene Aliases: CCDC56; COA3; HSPC009; MITRAC12
UniProt ID: (Human) Q9Y2R0
Entrez Gene ID: (Human) 28958
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.