Proteintech
Manufacturer Code:204271AP150UL
Catalog # 50-173-2884
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CCDC56 Fusion Protein Ag14260 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCDC56 HSPC009; coiled-coil domain containing 56; Coiled-coil domain-containing protein 56; Cytochrome c oxidase assembly factor 3 homolog, mitochondrial; cytochrome c oxidase assembly protein 3 homolog, mitochondrial; Mitochondrial translation regulation assembly intermediate of cytochrome c oxidase protein of 12 kDa
Gene Aliases: 1810033A19Rik; CCDC56; COA3; D11Ertd99e; HSPC009; MITRAC12; RGD1564337
UniProt ID: (Human) Q9Y2R0, (Mouse) Q9D2R6
Entrez Gene ID: (Human) 28958, (Rat) 498000, (Mouse) 52469
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.