Proteintech
Manufacturer Code:250491AP150UL
Catalog # 50-173-4693
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CCDC40 Fusion Protein Ag20034 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCDC40 KIAA1640; Coiled-coil domain-containing protein 40
Gene Aliases: CCDC40; CILD15; FAP172; KIAA1640
UniProt ID: (Human) A8MTD2
Entrez Gene ID: (Human) 55036
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.