Invitrogen
Manufacturer Code:MA527399
Catalog # PIMA527399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5C1 |
Immunogen | Full length human recombit protein of CCDC22 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CCDC22 Coiled-coil domain-containing protein 22 JM1 CXorf37; Coiled-coil domain-containing protein 22
Gene Aliases: CCDC22; CXorf37; JM1; RTSC2
UniProt ID: (Human) O60826
Entrez Gene ID: (Human) 28952
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.