Invitrogen
Manufacturer Code:PA551477
Catalog # PIPA551477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CCDC22 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCDC22 Coiled-coil domain-containing protein 22 JM1 CXorf37; ccdc22 {ECO:0000250|UniProtKB:O60826}; Coiled-coil domain-containing protein 22; coiled-coil domain-containing protein 22 {ECO:0000250|UniProtKB:O60826}
Gene Aliases: AI481216; CCDC22; CXorf37; DXImx40e; JM1; Ppp1r3f; RGD1560910; RTSC2; Sfc22
UniProt ID: (Human) O60826, (Mouse) Q9JIG7, (Rat) P86182
Entrez Gene ID: (Human) 28952, (Mouse) 54638, (Rat) 317381
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.