Novus Biologicals
Manufacturer Code:NBP191199
Catalog # NBP191199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ISRKHYKQIAQAAFNLKMRAACTGRTEYPKIRTFDGREHSTNSVNQDLLEAEKWTHIEGNVDIGDLTWEQKEKVLRLLFAKMNGCPSRKYNQSSRPPVPDYVVSDSGETKEFGDESKLQDKIFITQQIAISDSSGEVVLPTIPKEPQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basal body-orientation factor 1; C14orf45 chromosome 14 open reading frame 45 FLJ23189 hypothetical protein LOC80127; coiled-coil domain containing 176; Coiled-coil domain-containing protein 176
Gene Aliases: BBOF1; C14orf45; CCDC176
UniProt ID: (Human) Q8ND07
Entrez Gene ID: (Human) 80127
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.