Novus Biologicals
Manufacturer Code:NBP188164
Catalog # NBP188164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GSSASSAPQSVKRKAPDADDQDVKRAKATLVPQVQPSTSAWTTNFDKIGKEFIRATPSKPSGLSLLPDYEDEEEEEEEEEGDGER |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCDC16 coiled-coil domain containing 16 Coiled-coil domain-containing protein 16 MGC20398 OMCG1 SEL13 sel-13 suppressor of lin-12 homolog zinc finger protein 830; coiled-coil domain containing 16; Coiled-coil domain-containing protein 16; hypothetical protein MGC20398; orphan maintenance of genome 1; Zinc finger protein 830
Gene Aliases: CCDC16; OMCG1; ZNF830
UniProt ID: (Human) Q96NB3
Entrez Gene ID: (Human) 91603
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.