Invitrogen
Manufacturer Code:PA571685
Catalog # PIPA571685
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 291-320 amino acids from the C-terminal region of human C109B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium uniporter regulatory subunit MCUb mitochondrial MCUb Coiled-coil domain-containing protein 109B CCDC109B MCUB; Calcium uniporter regulatory subunit MCUb, mitochondrial; coiled-coil domain containing 109B; Coiled-coil domain-containing protein 109B; MCUb; mitochondrial calcium uniporter regulatory subunit MCUb
Gene Aliases: CCDC109B; MCUB
UniProt ID: (Human) Q9NWR8
Entrez Gene ID: (Human) 55013
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.