Invitrogen
Manufacturer Code:PA555338
Catalog # PIPA555338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CCBL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cysteine conjugate beta lyase 2; cysteine conjugate-beta lyase 2; Cysteine-S-conjugate beta-lyase 2; KATIII; Kynurenine aminotransferase 3; Kynurenine aminotransferase III; Kynurenine--glyoxylate transaminase; Kynurenine--oxoglutarate transaminase 3; Kynurenine--oxoglutarate transaminase III; kynurenine-oxoglutarate transaminase 3; RP11-82K18.3
Gene Aliases: CCBL2; KAT3; KATIII; KYAT3
UniProt ID: (Mouse) Q71RI9, (Rat) Q58FK9, (Human) Q6YP21
Entrez Gene ID: (Mouse) 229905, (Rat) 541589, (Human) 56267
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.