Abnova
Manufacturer Code:PAB21879
Catalog # 89-123-582
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human CCBL2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant CCBL2.
For Research Use Only
Protein Aliases: cysteine conjugate beta lyase 2; cysteine conjugate-beta lyase 2; Cysteine-S-conjugate beta-lyase 2; DKFZp547N1117; DKFZp667D0223; KAT3; KATIII; Kynurenine aminotransferase 3; Kynurenine aminotransferase III; Kynurenine--glyoxylate transaminase; Kynurenine--oxoglutarate transaminase 3; Kynurenine--oxoglutarate transaminase III; kynurenine-oxoglutarate transaminase 3; MGC9398; RBM1; RBMXL1; RP11-82K18.3; RP4-531M19.2
Gene Aliases: CCBL2; KAT3; KATIII; KYAT3
UniProt ID: (Human) Q6YP21
Entrez Gene ID: (Human) 56267
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.