Invitrogen
Manufacturer Code:PA555681
Catalog # PIPA555681
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CCBL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta-lysase, kidney; cysteine conjugate beta lyase 1; cysteine conjugate-beta lyase, cytoplasmic; cysteine conjugate-beta lyase; cytoplasmic (glutamine transaminase K, kyneurenine aminotransferase); Cysteine-S-conjugate beta-lyase; Glutamine transaminase K; Glutamine--phenylpyruvate transaminase; glutamine-phenylpyruvate aminotransferase; GTK; KATI; kyneurenine aminotransferase; Kynurenine aminotransferase 1; Kynurenine aminotransferase I; Kynurenine--oxoglutarate transaminase 1; Kynurenine--oxoglutarate transaminase 1 Cysteine-S-conjugate beta-lyase Glutamine transaminase K GTK Glutamine--phenylpyruvate transaminase Kynurenine aminotransferase I KATI Kynurenine--oxoglutarate transaminase I CCBL1; Kynurenine--oxoglutarate transaminase I
Gene Aliases: CCBL1; GTK; KAT1; KATI; KYAT1
UniProt ID: (Human) Q16773
Entrez Gene ID: (Human) 883
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.