Invitrogen
Manufacturer Code:PA5109485
Catalog # PIPA5109485
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human CCBL1 (NP_001116144.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010009K05Rik AI182306 Ccbl1 Cysteine-S-conjugate beta-lyase Glutamine transaminase K Glutamine--phenylpyruvate transaminase GTK Kat KAT1 KATI KYAT1 Kynurenine aminotransferase 1 Kynurenine aminotransferase I Kynurenine--oxoglutarate transaminase 1 Kynurenine--oxoglutarate transaminase 1 mitochondrial Kynurenine--oxoglutarate transaminase I; beta-lysase, kidney; cysteine conjugate beta lyase 1; cysteine conjugate-beta lyase 1; cysteine conjugate-beta lyase, cytoplasmic; cysteine conjugate-beta lyase; cytoplasmic (glutamine transaminase K, kyneurenine aminotransferase); Cysteine-S-conjugate beta-lyase; cytoplasmic (glutamine transaminase K, kyneurenine aminotransferase); Glutamine transaminase K; Glutamine--phenylpyruvate transaminase; glutamine-phenylpyruvate aminotransferase; GTK; KATI; kyneurenine aminotransferase; Kynurenine aminotransferase 1; Kynurenine aminotransferase I; kynurenine aminotransferase/glutamine transaminase K; Kynurenine--oxoglutarate transaminase 1; kynurenine--oxoglutarate transaminase 1, mitochondrial; Kynurenine--oxoglutarate transaminase I
Gene Aliases: 2010009K05Rik; AI182306; CCBL1; GTK; Kat; KAT1; KATI; KYAT1
UniProt ID: (Rat) Q08415, (Mouse) Q8BTY1, (Human) Q16773
Entrez Gene ID: (Rat) 311844, (Mouse) 70266, (Human) 883
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.