Abnova
Manufacturer Code:H00000883K
Catalog # 89-208-290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CCBL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CCBL1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: beta-lysase, kidney; cysteine conjugate beta lyase 1; cysteine conjugate-beta lyase, cytoplasmic; cysteine conjugate-beta lyase; cytoplasmic (glutamine transaminase K, kyneurenine aminotransferase); Cysteine-S-conjugate beta-lyase; FLJ95217 GTK KAT1 KATI MGC29624; Glutamine transaminase K; Glutamine--phenylpyruvate transaminase; glutamine-phenylpyruvate aminotransferase; GTK; KATI; kyneurenine aminotransferase; Kynurenine aminotransferase 1; Kynurenine aminotransferase I; Kynurenine--oxoglutarate transaminase 1; Kynurenine--oxoglutarate transaminase I
Gene Aliases: CCBL1; GTK; KAT1; KATI; KYAT1
UniProt ID: (Human) Q16773
Entrez Gene ID: (Human) 883
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.