Novus Biologicals
Manufacturer Code:NBP221998UV
Catalog # NBP221998UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 25G14 |
Immunogen | This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human CC chemokine receptor D6. Rat anti human CC Chemokine Receptor D6 O00590. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Atypical chemokine receptor 2; C-C chemokine receptor D6; C-C chemokine receptor D6 CCR10CCR9D6CC-chemokine-binding receptor JAB61 chemokine (C-C) receptor 9 chemokine binding protein 2 Chemokine receptor CCR-10 Chemokine receptor CCR-9 chemokine receptor D6 chemokine-binding protein 2 Chemokine-binding protein D6 CMKBR9chemokine (C-C motif) receptor 9 hD6 MGC126678 MGC138250; CC-chemokine-binding receptor JAB61; chemokine (C-C motif) receptor 9; chemokine (C-C) receptor 9; chemokine binding protein 2; Chemokine receptor CCR-10; Chemokine receptor CCR-9; chemokine receptor D6; Chemokine-binding protein 2; Chemokine-binding protein D6
Gene Aliases: ACKR2; CCBP2; CCR10; CCR9; CMKBR9; D6; hD6
UniProt ID: (Human) O00590
Entrez Gene ID: (Human) 1238
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.