Proteintech
Manufacturer Code:185441AP150UL
Catalog # 50-173-2178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CBX4 Fusion Protein Ag13425 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CBX4 Chromobox protein homolog 4 E3 SUMO protein ligase CBX4 hPC2 NBP16 PC2 Polycomb 2 homolog; chromobox homolog 4; chromobox homolog 4 (Pc class homolog, Drosophila); Chromobox protein homolog 4; chromobox-like protein 4; E3 SUMO-protein ligase CBX4; E3 SUMO-protein transferase CBX4; NS5ATP1-binding protein 16; Pc class 2 homolog; Pc2; Polycomb 2 homolog
Gene Aliases: CBX4; MPc2; NBP16; PC2
UniProt ID: (Human) O00257, (Mouse) O55187
Entrez Gene ID: (Human) 8535, (Mouse) 12418, (Rat) 501403
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.