Novus Biologicals
Manufacturer Code:NBP298758H
Catalog # NB055223
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human CBX3. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromobox homolog 3 (HP1 gamma homolog, Drosophila); chromobox homolog 3 chromobox homolog 3 (Drosophila HP1 gamma) chromobox protein homolog 3 Drosophila) HECH Heterochromatin protein 1 homolog gamma heterochromatin protein HP1 gamma heterochromatin-like protein 1 HP1 gamma HP1 gamma homolog HP1-GAMMA Modifier 2 protein; Chromobox protein homolog 3; HECH; Heterochromatin protein 1 homolog gamma; heterochromatin protein HP1 gamma; heterochromatin-like protein 1; HP1 gamma; HP1 gamma homolog; Modifier 2 protein
Gene Aliases: CBX3; HECH; HP1-GAMMA; HP1Hs-gamma
UniProt ID: (Human) Q13185
Entrez Gene ID: (Human) 11335
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.