Novus Biologicals
Manufacturer Code:NBP298756JF549
Catalog # NB105576
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human CBX1. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CBXM31HP1-BETA chromobox homolog 1 chromobox homolog 1 (Drosophila HP1 beta) chromobox homolog 1 (HP1 beta homolog Drosophila ) chromobox protein homolog 1 Heterochromatin protein 1 homolog beta heterochromatin protein 1-beta Heterochromatin protein p25 heterochromatin protein p25 beta HP1 beta HP1 beta homolog HP1Hsbeta HP1Hs-beta MOD1 Modifier 1 protein p25beta; chromobox homolog 1 (HP1 beta homolog Drosophila ); Chromobox protein homolog 1; Heterochromatin protein 1 homolog beta; heterochromatin protein 1-beta; Heterochromatin protein p25; heterochromatin protein p25 beta; HP1 beta; HP1 beta homolog; HP1Hsbeta; M31; Modifier 1 protein; p25beta
Gene Aliases: CBX; CBX1; HP1-BETA; HP1Hs-beta; HP1Hsbeta; M31; MOD1; p25beta
UniProt ID: (Human) P83916
Entrez Gene ID: (Human) 10951
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.