Novus Biologicals
Manufacturer Code:NBP243604
Catalog # NBP243604
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 519 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human CBS. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-thionase; Cystathionine beta-synthase; Cystathionine beta-synthase-like protein; cystathionine-beta-synthase EC 4.2.1.2210Beta-thionase; methylcysteine synthase; Serine sulfhydrase
Gene Aliases: CBS; CBSL; HIP4
UniProt ID: (Human) P35520
Entrez Gene ID: (Human) 875
Molecular Function:
deaminase
dehydratase
epimerase/racemase
hydrolase
isomerase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.