Invitrogen
Manufacturer Code:PA594923
Catalog # PIPA594923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human CBS recombit protein (Position: A331-K551). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-thionase; Cystathionine beta-synthase; Cystathionine beta-synthase 4.2.1.22 Beta-thionase Serine sulfhydrase CBS; Cystathionine beta-synthase-like protein; cystathionine-beta-synthase; Hemoprotein H-450; methylcysteine synthase; Serine sulfhydrase
Gene Aliases: CBS; CBSL; HIP4
UniProt ID: (Human) P35520, (Rat) P32232
Entrez Gene ID: (Human) 875, (Rat) 24250
Molecular Function: deaminase dehydratase epimerase/racemase hydrolase isomerase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.