Proteintech
Manufacturer Code:147871AP150UL
Catalog # 50-172-9721
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CBS Fusion Protein Ag6437 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta thionase CBS cystathionine beta synthase HIP4 Serine sulfhydrase; Beta-thionase; Cystathionine beta-synthase; Cystathionine beta-synthase-like protein; cystathionine-beta-synthase; Hemoprotein H-450; methylcysteine synthase; Serine sulfhydrase
Gene Aliases: AI047524; AI303044; CBS; CBSL; HIP4
UniProt ID: (Human) P35520, (Rat) P32232, (Mouse) Q91WT9
Entrez Gene ID: (Human) 875, (Rat) 24250, (Mouse) 12411
Molecular Function:
deaminase
dehydratase
epimerase/racemase
hydrolase
isomerase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.