Invitrogen
Manufacturer Code:MA525626
Catalog # PIMA525626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G6 |
Immunogen | Full length human recombit protein of CBR3 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: carbonyl reductase (NADPH) 3; Carbonyl reductase [NADPH] 3; epididymis secretory protein Li 25; NADPH-dependent carbonyl reductase 3; SDR21C2 hCBR3 NADPH-dependent carbonyl reductase 3 carbonyl reductase (NADPH) 3 carbonyl reductase &lsqbNADPH] 3 short chain dehydrogenase/reductase family 21C member 2; Short chain dehydrogenase/reductase family 21C member 2; short chain dehydrogenase/reductase family 21C, member 2
Gene Aliases: 1110001J05Rik; C81353; CBR3; hCBR3; HEL-S-25; SDR21C2
UniProt ID: (Human) O75828, (Mouse) Q8K354
Entrez Gene ID: (Dog) 487748, (Rhesus monkey) 695769, (Human) 874, (Mouse) 109857, (Rat) 304078
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.