Abnova
Manufacturer Code:H00000874K
Catalog # 89-205-335
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CBR3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CBR3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: carbonyl reductase (NADPH) 3; Carbonyl reductase [NADPH] 3; epididymis secretory protein Li 25; NADPH-dependent carbonyl reductase 3; SDR21C2 hCBR3; Short chain dehydrogenase/reductase family 21C member 2; short chain dehydrogenase/reductase family 21C, member 2
Gene Aliases: CBR3; hCBR3; HEL-S-25; SDR21C2
UniProt ID: (Human) O75828
Entrez Gene ID: (Human) 874
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.