Invitrogen
Manufacturer Code:PA5101494
Catalog # PIPA5101494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CBR1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase 15-hydroxyprostaglandin dehydrogenase [NADP(+)] 20-beta-hydroxysteroid dehydrogenase AW261796 carbonyl reductase (NADPH) 1 carbonyl reductase [NADPH] 1 Carbonyl reductase [NADPH] 1 (NADPH-dependent carbonyl reductase 1) carbonyl reductase 1 Cbr Cbr 1 CBR1 CR CRN hCBR1 NADPH-dependent carbonyl reductase 1 Prostaglandin 9-ketoreductase prostaglandin-E(2) 9-reductase SDR21C1 Short chain dehydrogenase/reductase family 21C member 1 short chain dehydrogenase/reductase family 21C member 1; 15-hydroxyprostaglandin dehydrogenase [NADP(+)]; 20-beta-hydroxysteroid dehydrogenase; carbonyl reductase (NADPH) 1; Carbonyl reductase [NADPH] 1; Carbonyl reductase [NADPH] 1 (NADPH-dependent carbonyl reductase 1); NADPH-dependent carbonyl reductase 1; PG-9-KR; Prostaglandin 9-ketoreductase; Prostaglandin-E(2) 9-reductase; Short chain dehydrogenase/reductase family 21C member 1; short chain dehydrogenase/reductase family 21C, member 1
Gene Aliases: AW261796; CBR; CBR1; CR; CRN; hCBR1; SDR21C1
UniProt ID: (Human) P16152, (Mouse) P48758
Entrez Gene ID: (Human) 873, (Mouse) 12408
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.