Invitrogen
Manufacturer Code:PA599492
Catalog # PIPA599492
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human P300/CBP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A430090G16 A730011L11 AW558298 CBP CBP/p300 CREB binding protein CREB-binding protein CREBBP E1A binding protein p300 E1A-associated protein p300 E1A-binding protein 300kD EP300 histone acetyltransferase p300 Histone butyryltransferase p300 Histone crotonyltransferase p300 KAT3A KAT3B p300 p300 HAT p300/CBP Protein propionyltransferase p300 RSTS RSTS2 RTS; CREB-binding protein; E1A-associated protein p300; E1A-binding protein, 300kD; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; Histone lysine acetyltransferase CREBBP; p300 HAT; Protein 2-hydroxyisobutyryltransferase p300; Protein lactyltransferas p300; Protein propionyltransferase p300; Protein-lysine acetyltransferase CREBBP
Gene Aliases: A430090G16; A730011L11; AW558298; CBP; CBP/p300; CREBBP; EP300; KAT3A; KAT3B; P300; p300 HAT; p300/CBP; RSTS; RSTS2; RTS
UniProt ID: (Human) Q92793, (Human) Q09472, (Mouse) B2RWS6
Entrez Gene ID: (Human) 1387, (Mouse) 12914, (Rat) 54244, (Human) 2033, (Mouse) 328572, (Rat) 170915
Molecular Function: DNA binding protein acetyltransferase chromatin/chromatin-binding protein nucleic acid binding transcription cofactor transcription factor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.