Invitrogen
Manufacturer Code:MA513634
Catalog # PIMA513634
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NM11 |
Immunogen | E1A affinity purified p300 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CREB-binding protein; Histone lysine acetyltransferase CREBBP; Protein-lysine acetyltransferase CREBBP
Gene Aliases: AW558298; CBP; CBP/p300; CREBBP; KAT3A; p300/CBP; RSTS; RTS
UniProt ID: (Human) Q92793
Entrez Gene ID: (Human) 1387, (Mouse) 12914, (Rat) 54244
Molecular Function:
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transcription cofactor
transcription factor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.