Proteintech
Manufacturer Code:205591AP150UL
Catalog # 50-173-2961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CBLN4 CBLNL1 Cerebellin 4 cerebellin 4 precursor Cerebellin like glycoprotein 1; cerebellin 4 precursor protein; cerebellin precursor-like 1; Cerebellin-4; Cerebellin-like glycoprotein 1
Gene Aliases: AI848962; CBLN4; CBLNL1; UNQ718/PRO1382
UniProt ID: (Human) Q9NTU7, (Mouse) Q8BME9
Entrez Gene ID: (Human) 140689, (Rat) 499947, (Mouse) 228942
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.