Proteintech
Manufacturer Code:211791AP150UL
Catalog # 50-173-3302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CBLL1 Fusion Protein Ag15463 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c Cbl like protein 1 CBLL1 HAKAI RING finger protein 188 RNF188; c-Cbl-like protein 1; Cas-Br-M (murine) ecotropic retroviral transforming sequence-like 1; Casitas B-lineage lymphoma-like 1; Casitas B-lineage lymphoma-transforming sequence-like protein 1; Cbl proto-oncogene like 1, E3 ubiquitin protein ligase; Cbl proto-oncogene, E3 ubiquitin protein ligase-like 1; Cbl proto-oncogene-like 1, E3 ubiquitin protein ligase; E-cadherin binding protein E7; E3 ubiquitin-protein ligase Hakai; RING finger protein 188; RING-type E3 ubiquitin transferase Hakai
Gene Aliases: AI467391; c-Cbl-like; CBLL1; HAKAI; RNF188
UniProt ID: (Human) Q75N03, (Mouse) Q9JIY2
Entrez Gene ID: (Human) 79872, (Mouse) 104836, (Rat) 314028
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.