Abnova
Manufacturer Code:MAB1380
Catalog # 89-110-396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M161 |
Immunogen | Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y700 of human CBL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic phosphopeptide of CBL.
For Research Use Only
Protein Aliases: C-CBL; Cas-Br-M (murine) ecotropic retroviral transforming sequence; Casitas B-lineage lymphoma proto-oncogene; Cbl proto-oncogene, E3 ubiquitin protein ligase; CBL2; E3 ubiquitin-protein ligase CBL; fragile site, folic acid type, rare, fra(11)(q23.3); oncogene CBL2; Proto-oncogene c-Cbl; RING finger protein 55; RING-type E3 ubiquitin transferase CBL; RNF55; Signal transduction protein CBL
Gene Aliases: C-CBL; CBL; CBL2; FRA11B; NSLL; RNF55
UniProt ID: (Human) P22681
Entrez Gene ID: (Human) 867
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.