Novus Biologicals
Manufacturer Code:NBP191753
Catalog # NBP191753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:DYSDRTISDEDESDEDMFMKFVSEDLHRCALLTADSFGDPFFPRTTQILLEYQLGRWVPRLREPRDLYGVSSSGPLSPTRWPYHCEVIDE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP/GTP binding protein-like 3; ATP/GTP binding protein-like 3 ATP/GTP-binding protein-like 3 CCP3 cytosolic carboxypeptidase 3 EC 3.4.17 EC 3.4.17.- FLJ12983 MGC32955; ATP/GTP-binding protein-like 3; Cytosolic carboxypeptidase 3
Gene Aliases: AGBL3; CCP3
UniProt ID: (Human) Q8NEM8
Entrez Gene ID: (Human) 340351
Molecular Function: G-protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.