Novus Biologicals
Manufacturer Code:NBP287129
Catalog # NB034751
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human CBARA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Allergen Hom s 4 ara CALC Atopy-related autoantigen CALC CALCDKFZp564C246 calcium binding atopy-related autoantigen 1 Calcium-binding atopy-related autoantigen 1 MICU1mitochondrial mitochondrial calcium uptake 1; ara CALC; Atopy-related autoantigen CALC; Calcium uptake protein 1, mitochondrial; Calcium-binding atopy-related autoantigen 1; Hom s 4
Gene Aliases: CALC; CBARA1; EFHA3; MICU1; MPXPS
UniProt ID: (Human) Q9BPX6
Entrez Gene ID: (Human) 10367
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.