Novus Biologicals
Manufacturer Code:NBP191751
Catalog # NBP191751
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GQQQRITFSETGAEEEEENDQLPLVHCVVARSEKSGLLQEPLAGGPLSNLSENTCDNFCLVLEAIQENLRQYKEIRD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cation channel sperm-associated protein 4; cation channel sperm-associated protein 4 cation channel sperm associated 4; cation channel, sperm associated 4; CatSper4; ion channel CatSper4
Gene Aliases: CATSPER4
UniProt ID: (Human) Q7RTX7
Entrez Gene ID: (Human) 378807
Molecular Function:
calcium channel
ion channel
sodium channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.