Invitrogen
Manufacturer Code:PA577359
Catalog # PIPA577359
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RALFQDSDPKRFQN corresponding to amino acid residues 508-521 of mouse CatSper1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cation channel of sperm 1; Cation channel sperm-associated protein 1; cation channel, sperm associated 1; CatSper1; sperm ion channel; sperm protein CATSPER1
Gene Aliases: Catsper; Catsper1; KSper
UniProt ID: (Mouse) Q91ZR5
Entrez Gene ID: (Mouse) 225865, (Rat) 689349
Molecular Function:
calcium channel
ion channel
sodium channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.