Invitrogen
Manufacturer Code:PA523743
Catalog # PIPA523743
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1574-1601 amino acids from the C-terminal region of human CASZ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: castor homolog 1, zinc finger; Castor-related protein; CST SRG ZNF693 dJ734G22.1 castor homolog 1 zinc finger castor-related protein putative survival-related protein survival related survival-related zinc finger protein 693 zinc finger protein castor homolog 1; Putative survival-related protein; survival-related; Zinc finger protein 693; Zinc finger protein castor homolog 1
Gene Aliases: 2410019P08Rik; AV096627; BC040081; CAS11; CASZ1; CST; D4Ertd432e; dJ734G22.1; Kiaa3026; SRG; ZNF693
UniProt ID: (Human) Q86V15, (Mouse) Q9CWL2
Entrez Gene ID: (Human) 54897, (Mouse) 69743
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.