Novus Biologicals
Manufacturer Code:NBP287125
Catalog # NB034747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human CASZ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: castor castor homolog 1 zinc finger castor homolog 1 zinc finger (Drosophila) castor zinc finger 1 Castor-related protein CST dJ734G22.1 FLJ20321 Putative survival-related protein SRG SRGcst survival related survival-related Zinc finger protein 693 zinc finger protein castor homolog 1 ZNF693 ZNF693FLJ12223; castor homolog 1, zinc finger; Castor-related protein; Putative survival-related protein; survival-related; Zinc finger protein 693; Zinc finger protein castor homolog 1
Gene Aliases: CAS11; CASZ1; CST; dJ734G22.1; SRG; ZNF693
UniProt ID: (Human) Q86V15
Entrez Gene ID: (Human) 54897
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.