Abnova
Manufacturer Code:H00054897B01P
Catalog # 89-335-015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | CASZ1 (ABZ92343.1 1 a.a. &sim 1044 a.a) full-length human protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Polyclonal Antibody.
Protein Aliases: castor homolog 1, zinc finger; Castor-related protein; CST FLJ12223 FLJ20321 SRG ZNF693 dJ734G22.1; Putative survival-related protein; survival-related; Zinc finger protein 693; Zinc finger protein castor homolog 1
Gene Aliases: CAS11; CASZ1; CST; dJ734G22.1; SRG; ZNF693
UniProt ID: (Human) Q86V15
Entrez Gene ID: (Human) 54897
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.