Abnova
Manufacturer Code:PAB10183
Catalog # 89-114-683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human CASZ1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CASZ1.
For Research Use Only
Protein Aliases: castor homolog 1, zinc finger; Castor-related protein; CST; dJ734G22.1; FLJ12223; FLJ20321; Putative survival-related protein; SRG; survival-related; Zinc finger protein 693; Zinc finger protein castor homolog 1; ZNF693
Gene Aliases: CAS11; CASZ1; CST; dJ734G22.1; SRG; ZNF693
UniProt ID: (Human) Q86V15
Entrez Gene ID: (Human) 54897
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.