Abnova
Manufacturer Code:H00008837M01
Catalog # 89-014-227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F12 |
Immunogen | CFLAR (AAH01602 178 a.a. ~ 277 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant CFLAR.
For Research Use Only
Protein Aliases: c-FLIP; c-FLIPL; c-FLIPR; c-FLIPS; CASH; CASP8 and FADD-like apoptosis regulator; CASP8 and FADD-like apoptosis regulator subunit p12; CASP8 and FADD-like apoptosis regulator subunit p43; CASP8AP1; Caspase homolog; Caspase-eight-related protein; Caspase-like apoptosis regulatory protein; caspase-related inducer of apoptosis; Casper; Cellular FLICE-like inhibitory protein; CLARP; FADD-like antiapoptotic molecule 1; FLAME; FLAME-1; FLAME1; FLIP; I-FLICE; Inhibitor of FLICE; MACH-related inducer of toxicity; MRIT; testis secretory sperm-binding protein Li 233m; Usurpin; usurpin beta
Gene Aliases: c-FLIP; c-FLIPL; c-FLIPR; c-FLIPS; CASH; CASP8AP1; Casper; CFLAR; CLARP; FLAME; FLAME-1; FLAME1; FLIP; I-FLICE; MRIT
UniProt ID: (Human) O15519
Entrez Gene ID: (Human) 8837
Molecular Function: cysteine protease enzyme modulator hydrolase protease protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.