Abnova
Manufacturer Code:PAB12873
Catalog # 89-116-150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 16 amino acids of human CASP14. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CASP14.
For Research Use Only
Protein Aliases: apoptosis-related cysteine protease; CASP-14; caspase 14, apoptosis-related cysteine peptidase; caspase 14, apoptosis-related cysteine protease; Caspase-14; Caspase-14 subunit p10, mature form; Caspase-14 subunit p17, mature form; Caspase-14 subunit p20, intermediate form; Caspase-14 subunit p8, intermediate form; MGC119078; MGC119079
Gene Aliases: CASP14
UniProt ID: (Human) P31944
Entrez Gene ID: (Human) 23581
Molecular Function:
cysteine protease
enzyme modulator
hydrolase
protease
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.