Invitrogen
Manufacturer Code:PA125080
Catalog # PIPA125080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH corresponding to residues K(393) S S P Q I R N P P S D A V Q R A(409) of human CASK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium/calmodulin-dependent serine protein kinase; G-protein-coupled receptor GPR34; Peripheral plasma membrane protein CASK; RP23-278L4.1 DXPri1 DXRib1 LIN-2 MGC7449 Pals3 mLin-2
Gene Aliases: Cask; DXPri1; DXRib1; LIN-2; mLin-2; Pals3
UniProt ID: (Rat) Q62915, (Mouse) O70589
Entrez Gene ID: (Dog) 480887, (Rat) 29647, (Mouse) 12361
Molecular Function:
cell junction protein
kinase
nucleotide kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.