Novus Biologicals
Manufacturer Code:NBP241181
Catalog # NBP241181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human CASK. The immunogen is located within amino acids 760 - 810 of CASK. Amino Acid Squence: YPIPHTTRPPKKDEENGKN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium/calmodulin-dependent serin protein kinase; Calcium/calmodulin-dependent serine protein kinase; calcium/calmodulin-dependent serine protein kinase (MAGUK family); Calcium/calmodulin-dependent serine protein kinase calcium/calmodulin-dependent serine protein kinase (MAGUK family) calcium/calmodulin-dependent serine protein kinase membrane-associatedguanylate kinase CAMGUK CMG EC 2.7.11 EC 2.7.11.1 FGS4CAGH39 FLJ22219 hCASK LIN2FLJ31914 MICPCH peripheral plasma membrane protein CASK Protein lin-2 homolog TNRC8 trinucleotide repeat containing 8; calcium/calmodulin-dependent serine protein kinase membrane-associated guanylate kinase; hCASK; Peripheral plasma membrane protein CASK; Protein lin-2 homolog; trinucleotide repeat containing 8
Gene Aliases: CAGH39; CAMGUK; CASK; CMG; FGS4; LIN2; MICPCH; MRXSNA; TNRC8
UniProt ID: (Human) O14936
Entrez Gene ID: (Human) 8573
Molecular Function: cell junction protein kinase nucleotide kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.