Proteintech
Manufacturer Code:552771AP150UL
Catalog # 50-173-6094
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAGH39 CASK CMG FGS4 hCASK LIN2 MICPCH Protein lin 2 homolog TNRC8; calcium/calmodulin-dependent serin protein kinase; Calcium/calmodulin-dependent serine protein kinase; calcium/calmodulin-dependent serine protein kinase (MAGUK family); calcium/calmodulin-dependent serine protein kinase membrane-associated guanylate kinase; hCASK; Peripheral plasma membrane protein CASK; Protein lin-2 homolog; trinucleotide repeat containing 8
Gene Aliases: CAGH39; CAMGUK; CASK; CMG; DXPri1; DXRib1; FGS4; LIN-2; LIN2; MICPCH; mLin-2; MRXSNA; Pals3; TNRC8
UniProt ID: (Human) O14936, (Rat) Q62915, (Mouse) O70589
Entrez Gene ID: (Human) 8573, (Rat) 29647, (Mouse) 12361
Molecular Function:
cell junction protein
kinase
nucleotide kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.